missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(2,5-difluorophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64517.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15431769

  • £398.56 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Chloro-2-(2,5-difluorophenyl)benzimidazole
264.66
MFCD20543594

Specifications

5-Chloro-2-(2,5-difluorophenyl)benzimidazole
Solid
264.66
5g
95%
MFCD20543594
Videos
SDS