missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-2701447
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175203-78-8
C6H2Cl2N2O3S
253.05
MFCD00203878
ZFBZICACMDZKNQ-UHFFFAOYSA-N
5-chlorobenzofurazan-4-sulfonyl chloride, 5-chlorobenzo c 1,2,5 oxadiazole-4-sulfonyl chloride, pubchem5077, 2,1,3-benzoxadiazole-4-sulfonylchloride,5-chloro, 5-chloro-2,1,3-benzoxadiazole-4-sulphonyl chloride, 2,1,3-benzoxadiazole-4-sulfonylchloride, 5-chloro, 5-chloranyl-2,1,3-benzoxadiazole-4-sulfonyl chloride, 5-chloro-benzo 1,2,5 oxadiazole-4-sulfonyl chloride
2779275
5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride
ClC1=C(C2=NON=C2C=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
175203-78-8
C6H2Cl2N2O3S
253.05
MFCD00203878
ZFBZICACMDZKNQ-UHFFFAOYSA-N
5-chlorobenzofurazan-4-sulfonyl chloride, 5-chlorobenzo c 1,2,5 oxadiazole-4-sulfonyl chloride, pubchem5077, 2,1,3-benzoxadiazole-4-sulfonylchloride,5-chloro, 5-chloro-2,1,3-benzoxadiazole-4-sulphonyl chloride, 2,1,3-benzoxadiazole-4-sulfonylchloride, 5-chloro, 5-chloranyl-2,1,3-benzoxadiazole-4-sulfonyl chloride, 5-chloro-benzo 1,2,5 oxadiazole-4-sulfonyl chloride
2779275
5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride
ClC1=C(C2=NON=C2C=C1)S(Cl)(=O)=O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 253.06
Percent Purity 97%
Chemical Name or Material 5-chloro-2, 1, 3-benzoxadiazole-4-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'