missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 228.56 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'