missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde, ≥95%, Maybridge

Brand:  Maybridge GK01024EA

Code : 31

Additional Details : CAS Number : 128455-62-9 Weight : 0.06000kg

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10562472

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
96.5
97%
C6H4ClF3N2O
MFCD00068130
128455-62-9
100.0
97%
212.56

Specifications

5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
96.5
Amber Glass Bottle
97%
C6H4ClF3N2O
MFCD00068130
128455-62-9
100.0
10g
97%
212.56
Videos
SDS