missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-chloro-1-benzothiophene-3-carbonitrile, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 193.66 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-1-benzothiophene-3-carbonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'