missing translation for 'onlineSavingsMsg'
Learn More

(5-Chloro-1-benzothiophen-3-yl)methylamine, 97%, Maybridge

Brand:  Maybridge SP01477DA

Code : 31

Additional Details : CAS Number : 71625-90-6 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10592083

  • £211.00 / 1g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

(5-Chloro-1-benzothiophen-3-yl)methylamine hydrochloride
97.0
97%
234.15
71625-90-6
97%
C9H8ClNS·ClH
MFCD01314327

Specifications

(5-Chloro-1-benzothiophen-3-yl)methylamine hydrochloride
71625-90-6
Amber Glass Bottle
97%
234.15
1g
97.0
97%
C9H8ClNS·ClH
MFCD01314327
Videos
SDS