missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-chloro-1,3-dimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
Synonym | 5-chloro-1,3-dimethyl-1h-pyrazole-4-carboxylic acid, 5-chloro-1,3-dimethyl-4-pyrazolecarboxylic acid, pubchem23627, chlorodimethylpyrazolecarboxylicacid, 4-carboxy-5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid,5-chloro-1,3-dimethyl, 1h-pyrazole-4-carboxylic acid, 5-chloro-1,3-dimethyl, 1h-pyrazole-4-carboxylicacid, 5-chloro-1,3-dimethyl |
Formula Weight | 174.59 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-1, 3-dimethyl-1H-pyrazole-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'