missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromothiophene-2-sulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
1g;
Specifications
CAS | 55854-46-1 |
Assay Percent Range | 97% |
Molecular Formula | C4H2BrClO2S2 |
MDL Number | MFCD00084909 |
Quantity | 1g |
Synonym | 5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride |
InChI Key | WGYBIEOLAFYDEC-UHFFFAOYSA-N |
SMILES | ClS(=O)(=O)C1=CC=C(Br)S1 |
IUPAC Name | 5-bromothiophene-2-sulfonyl chloride |
Molecular Weight (g/mol) | 261.53 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'