missing translation for 'onlineSavingsMsg'
Learn More

5-Bromosalicylic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10711821
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
89-55-4
C7H5BrO3
217.02
MFCD00002455
IEJOONSLOGAXNO-UHFFFAOYSA-N
5-bromosalicylic acid, benzoic acid, 5-bromo-2-hydroxy, salicylic acid, 5-bromo, 2-hydroxy-5-bromobenzoic acid, benzoic acid, 3-bromo-6-hydroxy, 5-bromo-2-hydroxy-benzoic acid, 5-bromo-2-hydroxybenzoic acid, 5-bromsalicylic acid, 5-bromosalicyclic acid, acmc-209r1n
6972
5-bromo-2-hydroxybenzoic acid
OC(=O)C1=CC(Br)=CC=C1O
This item is not returnable. View return policy
89-55-4
C7H5BrO3
217.02
MFCD00002455
IEJOONSLOGAXNO-UHFFFAOYSA-N
5-bromosalicylic acid, benzoic acid, 5-bromo-2-hydroxy, salicylic acid, 5-bromo, 2-hydroxy-5-bromobenzoic acid, benzoic acid, 3-bromo-6-hydroxy, 5-bromo-2-hydroxy-benzoic acid, 5-bromo-2-hydroxybenzoic acid, 5-bromsalicylic acid, 5-bromosalicyclic acid, acmc-209r1n
6972
5-bromo-2-hydroxybenzoic acid
OC(=O)C1=CC(Br)=CC=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 162.0°C to 166.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC6H3(OH)CO2H
Quantity 50 g
Beilstein 10, 107
Formula Weight 217.02
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromosalicylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromosalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'