missing translation for 'onlineSavingsMsg'
Learn More

5-Bromooxindole, 97%, Thermo Scientific™

Catalog Number p-4510407
Click to view available options
Quantity:
1 g
5 g
100 g
This item is not returnable. View return policy
20870-78-4
C8H6BrNO
212.046
VIMNAEVMZXIKFL-UHFFFAOYSA-N
5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
611193
5-bromo-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)Br)NC1=O
This item is not returnable. View return policy
20870-78-4
C8H6BrNO
212.046
VIMNAEVMZXIKFL-UHFFFAOYSA-N
5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
611193
5-bromo-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)Br)NC1=O
Assay Percent Range 97%
Quantity 1 g
Formula Weight 212.05
Percent Purity 97%
Chemical Name or Material 5-Bromo-1, 3-dihydro-2H-indol-2-one
compliance-icons
Product Identifier
  • 5-Bromooxindole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'