missing translation for 'onlineSavingsMsg'
Learn More

5-Bromooxindole, 97%, Thermo Scientific Chemicals

Catalog Number p-3777693
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20870-78-4
C8H6BrNO
212.05
MFCD00456998
VIMNAEVMZXIKFL-UHFFFAOYSA-N
5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
611193
5-bromo-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)Br)NC1=O
This item is not returnable. View return policy
20870-78-4
C8H6BrNO
212.05
MFCD00456998
VIMNAEVMZXIKFL-UHFFFAOYSA-N
5-bromooxindole, 5-bromoindolin-2-one, 5-bromo-2-oxindole, 5-bromo-1,3-dihydro-2h-indol-2-one, 5-bromo-2,3-dihydro-1h-indol-2-one, 5-bromo-2-indolinone, 5-bromo-1,3-dihydro-indol-2-one, 2h-indol-2-one, 5-bromo-1,3-dihydro, 5-bromo-2-oxindol, 1,3-dihydro-5-bromoindol-2-one
611193
5-bromo-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)Br)NC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 219.0°C to 221.0°C
Color Golden to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 212.05
Percent Purity 97%
Physical Form Crystals, Powder or Granules
Chemical Name or Material 5-Bromooxindole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromooxindole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'