Learn More
Thermo Scientific Maybridge 5-(Bromomethyl)-3-methylisoxazole, 1g, 95%
Brand: Thermo Scientific Maybridge CC27008DA
5-(Bromomethyl)-3-methylisoxazole, 1g, 95%
Code : 31
Additional Details : CAS Number : 36958-61-9 Weight : 0.05100kg Transport : UN number : 3265 Chem class : 8 Pack group : III