missing translation for 'onlineSavingsMsg'
Learn More

5-(bromomethyl)-3-methylisoxazole, 95%, Thermo Scientific™

Catalog Number p-3586796
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
36958-61-9
C5H6BrNO
176.013
MFCD09879901
DMUJLHLWCUMHRT-UHFFFAOYSA-N
5-bromomethyl-3-methylisoxazole, 5-bromomethyl-3-methyl-1,2-oxazole, isoxazole, 5-bromomethyl-3-methyl, 3-methyl-5-bromomethylisoxazole
10607354
5-(bromomethyl)-3-methyl-1,2-oxazole
CC1=NOC(=C1)CBr
This item is not returnable. View return policy
36958-61-9
C5H6BrNO
176.013
MFCD09879901
DMUJLHLWCUMHRT-UHFFFAOYSA-N
5-bromomethyl-3-methylisoxazole, 5-bromomethyl-3-methyl-1,2-oxazole, isoxazole, 5-bromomethyl-3-methyl, 3-methyl-5-bromomethylisoxazole
10607354
5-(bromomethyl)-3-methyl-1,2-oxazole
CC1=NOC(=C1)CBr
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.01
Percent Purity 95%
Chemical Name or Material 5-(Bromomethyl)-3-methylisoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'