missing translation for 'onlineSavingsMsg'
Learn More

5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15467386
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1256360-47-0
C13H17BBrFO2
314.989
MFCD17926519
UEBRMJPFPXLIMK-UHFFFAOYSA-N
2-5-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-bromomethyl-2-fluorophenylboronic acid pinacol ester, 5-bromomethyl-2-fluorophenylboronic acid, pinacol ester
53217273
2-[5-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)CBr)F
This item is not returnable. View return policy
1256360-47-0
C13H17BBrFO2
314.989
MFCD17926519
UEBRMJPFPXLIMK-UHFFFAOYSA-N
2-5-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-bromomethyl-2-fluorophenylboronic acid pinacol ester, 5-bromomethyl-2-fluorophenylboronic acid, pinacol ester
53217273
2-[5-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)CBr)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 314.99
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'