missing translation for 'onlineSavingsMsg'
Learn More

5-(Bromomethyl)-1-methyl-1H-1,2,3-benzotriazole, 90+%, Thermo Scientific™

Catalog Number p-4510257
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
499770-76-2
C8H8BrN3
226.077
OSUZHHPMRAIJDY-UHFFFAOYSA-N
5-bromomethyl-1-methyl-1h-benzo d 1,2,3 triazole, 5-bromomethyl-1-methyl-1h-1,2,3-benzotriazole, 5-bromomethyl-1-methylbenzotriazole, 5-bromomethyl-1-methyl-benzotriazole, 1h-benzotriazole, 5-bromomethyl-1-methyl, 5-bromomethyl-1-methyl-1,2,3-benzotriazole, 5-bromomethyl-1-methyl-1h-benzotriazole
2795418
5-(bromomethyl)-1-methylbenzotriazole
CN1C2=C(C=C(C=C2)CBr)N=N1
This item is not returnable. View return policy
499770-76-2
C8H8BrN3
226.077
OSUZHHPMRAIJDY-UHFFFAOYSA-N
5-bromomethyl-1-methyl-1h-benzo d 1,2,3 triazole, 5-bromomethyl-1-methyl-1h-1,2,3-benzotriazole, 5-bromomethyl-1-methylbenzotriazole, 5-bromomethyl-1-methyl-benzotriazole, 1h-benzotriazole, 5-bromomethyl-1-methyl, 5-bromomethyl-1-methyl-1,2,3-benzotriazole, 5-bromomethyl-1-methyl-1h-benzotriazole
2795418
5-(bromomethyl)-1-methylbenzotriazole
CN1C2=C(C=C(C=C2)CBr)N=N1
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 226.08
Percent Purity 95%
Chemical Name or Material 5-(Bromomethyl)-1-methyl-1H-1, 2, 3-benzotriazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'