missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzoxazole-2-thiol, 97%, Thermo Scientific™

Catalog Number 15414937
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 439607-87-1
Melting Point 280°C to 283°C
Molecular Formula C7H4BrNOS
MDL Number MFCD08447020,MFCD22556198
Quantity 1 g
Sensitivity Light and moisture sensitive
Synonym 5-bromobenzo d oxazole-2-thiol, 5-bromobenzoxazole-2-thiol, 5-bromobenzo d oxazole-2 3h-thione, 5-bromo-1,3-benzoxazole-2-thiol, acmc-20am5b, 5-bromo-2 3h-benzoxazolethione, 5-bromo-1,3-benzoxazole-2 3h-thione
InChI Key YFFBNUPHVZYFQL-UHFFFAOYSA-N
SMILES BrC1=CC=C2OC(=S)NC2=C1
IUPAC Name 5-bromo-3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol) 230.08
PubChem CID 10633197
Formula Weight 230.08
Percent Purity 97%
Chemical Name or Material 5-Bromobenzoxazole-2-thiol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'