missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzo[b]thiophene-3-carbaldehyde, 97%, Maybridge

Brand:  Maybridge CC33504DE

Code : 31

Additional Details : CAS Number : 16296-72-3 Weight : 0.05500kg

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10285274

  • £462.00 / 5g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Bromobenzo[b]thiophene-3-carbaldehyde
97%
241.11
97%
C9H5BrOS

Specifications

5-Bromobenzo[b]thiophene-3-carbaldehyde
Amber Glass Bottle
97%
C9H5BrOS
16296-72-3
5g
97%
241.11
Videos
SDS