missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzo[b]furan-2-carbonyl chloride, 97%, Thermo Scientific™

Catalog Number 10045762
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
62878-96-0
C9H4BrClO2
259.483
FGPAEHRRVKLUHA-UHFFFAOYSA-N
5-bromobenzo b furan-2-carbonyl chloride, 5-bromobenzofuran-2-carbonyl chloride
2795534
5-bromo-1-benzofuran-2-carbonyl chloride
C1=CC2=C(C=C1Br)C=C(O2)C(=O)Cl
This item is not returnable. View return policy
62878-96-0
C9H4BrClO2
259.483
FGPAEHRRVKLUHA-UHFFFAOYSA-N
5-bromobenzo b furan-2-carbonyl chloride, 5-bromobenzofuran-2-carbonyl chloride
2795534
5-bromo-1-benzofuran-2-carbonyl chloride
C1=CC2=C(C=C1Br)C=C(O2)C(=O)Cl
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 259.49
Percent Purity 95%
Chemical Name or Material 5-Bromobenzo[b]furan-2-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'