missing translation for 'onlineSavingsMsg'
Learn More

5-(Bromoacetyl)-2-oxoindoline, 97%, Thermo Scientific™

Catalog Number 10462284
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
105316-98-1
C10H8BrNO2
254.083
MFCD08690294
WHLZVVMOQHTDAX-UHFFFAOYSA-N
5-bromoacetyl-2-oxoindoline, 5-2-bromoacetyl indolin-2-one, 5-bromoacetyl-1,3-dihydro-2h-indol-2-one, 5-2-bromoacetyl-1,3-dihydroindol-2-one, 2h-indol-2-one,5-2-bromoacetyl-1,3-dihydro, 5-bromoacetyloxindole, acmc-20a59p, 5-alpha-bromoacetyl oxindole, 5-2-bromoacetyl-2-indolinone, 2h-indol-2-one, 5-bromoacetyl-1,3-dihydro
22099309
5-(2-bromoacetyl)-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)C(=O)CBr)NC1=O
This item is not returnable. View return policy
105316-98-1
C10H8BrNO2
254.083
MFCD08690294
WHLZVVMOQHTDAX-UHFFFAOYSA-N
5-bromoacetyl-2-oxoindoline, 5-2-bromoacetyl indolin-2-one, 5-bromoacetyl-1,3-dihydro-2h-indol-2-one, 5-2-bromoacetyl-1,3-dihydroindol-2-one, 2h-indol-2-one,5-2-bromoacetyl-1,3-dihydro, 5-bromoacetyloxindole, acmc-20a59p, 5-alpha-bromoacetyl oxindole, 5-2-bromoacetyl-2-indolinone, 2h-indol-2-one, 5-bromoacetyl-1,3-dihydro
22099309
5-(2-bromoacetyl)-1,3-dihydroindol-2-one
C1C2=C(C=CC(=C2)C(=O)CBr)NC1=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 254.08
Percent Purity 97%
Chemical Name or Material 5-(Bromoacetyl)-2-oxoindoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'