Learn More
5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals
Description
5-Bromo-5-nitro-1,3-dioxane is used as a stabilizer and preserving agent for biological molecules and solutions such as antibodies and antisera. Bronidox is used in a variety of rinse-off cosmetic. It can be used alone or in combination with methylisothiazolone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• 5-Bromo-5-nitro-1,3-dioxane is a chemical compound showing antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi
• This compound is capable of promoting the oxidation of essential protein thiols, causing inhibition of enzyme activity leading to the inhibition of microbial growth
Application
• 5-Bromo-5-nitro-1,3-dioxane has been used as a stabilizer and preserving agent for biological molecules and solutions, including antibodies and antisera
• It can be used alone or combined with methylisothiazolinone, which is also considered to be an effective preservative
Spécification
Melting Point | 58°C to 61°C |
Color | White |
Quantity | 5 g |
Solubility Information | Soluble in water at 12.5mg/ml |
Formula Weight | 212 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 5-Bromo-5-nitro-1,3-dioxane |
RUO – Research Use Only