Learn More
5-Bromo-4-methyl-2-phenyl-1,3-thiazole, ≥97%, Maybridge
Brand: Maybridge CC29210EA
Code : 31
Additional Details : CAS Number : 28771-82-6 Weight : 0.06000kg
Quantity | 10g |
---|---|
Packaging | Amber Glass Bottle |
Chemical Identifiers
5-Bromo-4-methyl-2-phenyl-1, 3-thiazole | |
96.5 | |
97% | |
C10H8BrNS | |
MFCD04974049 |
28771-82-6 | |
100.0 | |
97% | |
254.15 | |
5-Bromo-4-methyl-2-phenyl-1, 3-thiazole (IUPAC) |
Specifications
5-Bromo-4-methyl-2-phenyl-1, 3-thiazole | |
96.5 | |
Amber Glass Bottle | |
97% | |
C10H8BrNS | |
MFCD04974049 |
28771-82-6 | |
100.0 | |
10g | |
97% | |
254.15 | |
5-Bromo-4-methyl-2-phenyl-1, 3-thiazole (IUPAC) |