missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole, ≥97%, Thermo Scientific™

Catalog Number p-3075781
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
850375-27-8
C11H7BrF3NS
322.143
MFCD04974053
BZCIYGNYGIKEEC-UHFFFAOYSA-N
5-bromo-4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole, 5-bromo-4-methyl-2-4-trifluoromethyl-phenyl-1,3-thiazole, 5-bromo-4-methyl-2-4-trifluoromethyl phenyl thiazole, 5-bromanyl-4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole
2824063
5-bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)Br
This item is not returnable. View return policy
850375-27-8
C11H7BrF3NS
322.143
MFCD04974053
BZCIYGNYGIKEEC-UHFFFAOYSA-N
5-bromo-4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole, 5-bromo-4-methyl-2-4-trifluoromethyl-phenyl-1,3-thiazole, 5-bromo-4-methyl-2-4-trifluoromethyl phenyl thiazole, 5-bromanyl-4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole
2824063
5-bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)Br
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 322.15
Percent Purity 97%
Chemical Name or Material 5-Bromo-4-methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'