missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-methoxythiophene-3-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 10475312
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
162848-23-9
C6H5BrO3S
237.07
MFCD00068031
OMNYIQIDPSRNES-UHFFFAOYSA-N
3-thiophenecarboxylicacid, 5-bromo-4-methoxy, maybridge3_003417, 2-bromo-3-methoxythiophene-4-carboxylic acid, 5-bromo-4-methoxy-3-thiophenecarboxylic acid, 5-bromanyl-4-methoxy-thiophene-3-carboxylic acid
2778393
5-bromo-4-methoxythiophene-3-carboxylic acid
COC1=C(Br)SC=C1C(O)=O
This item is not returnable. View return policy
162848-23-9
C6H5BrO3S
237.07
MFCD00068031
OMNYIQIDPSRNES-UHFFFAOYSA-N
3-thiophenecarboxylicacid, 5-bromo-4-methoxy, maybridge3_003417, 2-bromo-3-methoxythiophene-4-carboxylic acid, 5-bromo-4-methoxy-3-thiophenecarboxylic acid, 5-bromanyl-4-methoxy-thiophene-3-carboxylic acid
2778393
5-bromo-4-methoxythiophene-3-carboxylic acid
COC1=C(Br)SC=C1C(O)=O
CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-4-methoxythiophene-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'