5-Bromo-4-chloro-3-indolyl sulfate potassium salt, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 337230500

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 6578-07-0 Weight: 0.05005kg


Packaging Glass bottle
Quantity 50mg

Product Code. 10164111

Quantity Price
1 £ 41.7 / 50mg
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 5-Bromo-4-chloro-3-indolyl sulfate potassium salt
Name Note 99%
CAS 6578-07-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H4BrClKNO4S
Formula Weight 364.63
MDL Number MFCD00056929
Synonym X-Sulfate potassium salt

Specification

Assay ≥98.5%
Physical Form Powder
Formula Weight 364.63
Quantity 50mg
Color Almost white to light yellow
CAS Min % 98.5
Infrared Spectrum Authentic
CAS 6578-07-0
Assay Percent Range 99%
MDL Number MFCD00056929
Packaging Glass bottle
Molecular Formula C8H4BrClKNO4S
CAS Max % 100.0
Name Note 99%
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl sulfate potassium salt
Synonym X-Sulfate potassium salt
Documentation

Technical documents