Learn More
Thermo Scientific Maybridge (5-Bromo-3-pyridinyl)methylamine, 5g, 97%
Brand: Thermo Scientific Maybridge CC67113DE
(5-Bromo-3-pyridinyl)methylamine, 5g, 97%
Code : 31
Additional Details : CAS Number : 135124-70-8 Weight : 0.05500kg Transport : UN number : 2922 Chem class : 8 Pack group : III