missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-3-methylpyridin-2-amine, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 187.04 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-3-methylpyridin-2-amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'