missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine, 97%, Acros Organics

Brand:  Acros Organics 452570010

Code : NEW

Additional Details : CAS Number : 1262412-30-5 Weight : 0.00100kg


Quantity 1g

 View more versions of this product

Product Code. 15209806

  • £158.80 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket


Specifications

Chemical Identifiers

5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine
97%
C6H2BrF4N
243.98

Specifications

5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine
97%
C6H2BrF4N
1262412-30-5
1g
243.98
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.