5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine, 97%, Acros Organics


Overview
Brand: Acros Organics

Manufacturer Part Number: 452570050

5GR 5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine,97%

UNSPSC: 12000000

Code: NEW

Additional Details:
Additional Details: CAS Number: 1262412-30-5 Weight: 0.01000kg


Quantity 5g

Product Code. 15219806

Quantity Price
1 £ 435.9 / 5g
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine
Assay Percent Range 97%
Molecular Formula C6H2BrF4N
Formula Weight 243.98

Specification

Formula Weight 243.98
Quantity 5g
Boiling Point 68.0° to 70.0°C (25.0 Torr)
CAS 1262412-30-5
Assay Percent Range 97%
Molecular Formula C6H2BrF4N
Chemical Name or Material 5-Bromo-3-fluoro-2-(trifluoromethyl)pyridine