missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-3-(chloromethyl)-1-benzothiophene, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 5-bromo-3-chloromethyl-1-benzothiophene, 5-bromo-3-chloromethyl benzo b thiophene, 5-bromo-3-chloromethyl-benzo b thiophene, benzo b thiophene,5-bromo-3-chloromethyl, benzo b thiophene, 5-bromo-3-chloromethyl, 5-bromo-3-chloromethyl-1-benzo b thiophene |
Formula Weight | 261.57 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-3-(chloromethyl)-1-benzothiophene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'