missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-chloro-2-methoxypyridine, 97%, Thermo Scientific™

Catalog Number p-4813201
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
848366-28-9
C6H5BrClNO
222.47
XCHXNAQRMMXBGN-UHFFFAOYSA-N
40427004
5-bromo-3-chloro-2-methoxypyridine
COC1=NC=C(C=C1Cl)Br
This item is not returnable. View return policy
848366-28-9
C6H5BrClNO
222.47
XCHXNAQRMMXBGN-UHFFFAOYSA-N
40427004
5-bromo-3-chloro-2-methoxypyridine
COC1=NC=C(C=C1Cl)Br
Melting Point 61°C to 63°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Synonym 2-methoxy-3-chloro-5-bromopyridine, 5-bromo-3-chloro-2-methoxy-pyridine, pyridine, 5-bromo-3-chloro-2-methoxy, 2-methoxyl-3-chloro-5bromopyridine, 3-chloro-2-methoxy-5-bromopyridine, pubchem1164, acmc-209pww, abbypharma ap-11-5379, 2-methoxy-3-chloro-5-bromo-yridine, pyridine,5-bromo-3-chloro-2-methoxy
Formula Weight 222.47
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-chloro-2-methoxypyridine
compliance-icons
Product Identifier
  • 5-Bromo-3-chloro-2-methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'