missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-3-chloro-2-methoxypyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
Melting Point | 61°C to 63°C |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Quantity | 5 g |
Synonym | 2-methoxy-3-chloro-5-bromopyridine, 5-bromo-3-chloro-2-methoxy-pyridine, pyridine, 5-bromo-3-chloro-2-methoxy, 2-methoxyl-3-chloro-5bromopyridine, 3-chloro-2-methoxy-5-bromopyridine, pubchem1164, acmc-209pww, abbypharma ap-11-5379, 2-methoxy-3-chloro-5-bromo-yridine, pyridine,5-bromo-3-chloro-2-methoxy |
Formula Weight | 222.47 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-3-chloro-2-methoxypyridine |
Safety and Handling
Product Identifier
- 5-Bromo-3-chloro-2-methoxypyridine
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'