missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-3-(4-methyl-1-piperazinylmethyl)indole, 95%

Brand:  Alfa Aesar™ H63023.03

Code : NEW

Additional Details : CAS Number : 701205-13-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458096

  • £95.61 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Bromo-3-(4-methyl-1-piperazinylmethyl)indole
308.22
11950923
701205-13-2
MFCD28126773

Specifications

5-Bromo-3-(4-methyl-1-piperazinylmethyl)indole
Solid
95%
MFCD28126773
701205-13-2
1g
308.22
11950923
Videos
SDS