missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-3-(4-methyl-1-piperazinylmethyl)indole, 95%

Brand:  Alfa Aesar™ H63023.06

Code : NEW

Additional Details : CAS Number : 701205-13-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15468096

  • £478.06 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Bromo-3-(4-methyl-1-piperazinylmethyl)indole
308.22
11950923
701205-13-2
MFCD28126773

Specifications

5-Bromo-3-(4-methyl-1-piperazinylmethyl)indole
Solid
95%
MFCD28126773
701205-13-2
5g
308.22
11950923
Videos
SDS