5-Bromo-3,4-dimethylbenzene-1,2-diamine, ≥90%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: BTB06763DA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 107100-16-3 Weight: 0.05100kg Transport: UN number : 2811 Chem class : 1 Pack group : III


Packaging Amber Glass Bottle
Quantity 1g

Product Code. 10364412

Quantity Price
1 £ 21.0 / 1g
Estimated Shipment date
from Supplier 28-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 5-Bromo-3, 4-dimethylbenzene-1, 2-diamine
CAS 107100-16-3
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C8H11BrN2
Formula Weight 215.09
MDL Number MFCD00052696

Specification

Assay 90%
Formula Weight 215.09
Quantity 1g
MDL Number MFCD00052696
Assay Percent Range 90%
Packaging Amber Glass Bottle
CAS 107100-16-3
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 5-Bromo-3, 4-dimethylbenzene-1, 2-diamine
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Molecular Formula C8H11BrN2
Health Hazard 1 GHS Signal Word: Warning
CAS Max % 100.0
CAS Min % 89.5
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.