missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-(3-oxycyclopentyl)indole, 95%, Thermo Scientific™

Catalog Number 15470609
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
676170-02-8
C13H12BrNO
278.149
OGXFMBGFSFOQGH-UHFFFAOYSA-N
3-5-bromo-1h-indol-3-yl cyclopentanone, 3-5-bromo-1h-indole-3-yl cyclopentanone, 3-5-bromo-1h-indol-3-yl cyclopentan-1-one
22139469
3-(5-bromo-1H-indol-3-yl)cyclopentan-1-one
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)Br
This item is not returnable. View return policy
676170-02-8
C13H12BrNO
278.149
OGXFMBGFSFOQGH-UHFFFAOYSA-N
3-5-bromo-1h-indol-3-yl cyclopentanone, 3-5-bromo-1h-indole-3-yl cyclopentanone, 3-5-bromo-1h-indol-3-yl cyclopentan-1-one
22139469
3-(5-bromo-1H-indol-3-yl)cyclopentan-1-one
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 278.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Bromo-3-(3-oxycyclopentyl)indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'