missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-3-(1-piperidinylmethyl)indole, 95%

Brand:  Alfa Aesar™ H63087.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479578

  • £95.61 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Bromo-3-(1-piperidinylmethyl)indole
293.2
MFCD28126772

Specifications

5-Bromo-3-(1-piperidinylmethyl)indole
Solid
293.2
1g
95%
MFCD28126772
Videos
SDS