missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole, 97%

Brand:  Alfa Aesar™ H63590.06

Code : NEW

Additional Details : CAS Number : 282546-99-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15408396

  • £298.92 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
334.21
14255489
282546-99-0
MFCD06411613
5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole

Specifications

5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
Solid
5g
MFCD06411613
5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)indole
282546-99-0
97%
334.21
14255489
Videos
SDS