Alfa Aesar™ 5-Bromo-2-(trifluoromethoxy)aniline, 98%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H61663.14

UNSPSC: 12352116

Code: NEW

Additional Details:
Additional Details: CAS Number: 886762-08-9 Weight: 0.02500kg


Quantity 25g

Product Code. 15478218

Quantity Price
1 £ 42.82 / 25g
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 5-Bromo-2-(trifluoromethoxy)aniline
CAS 886762-08-9
Formula Weight 256.02
MDL Number MFCD04039242
UN Number UN2810

Specification

Formula Weight 256.02
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD04039242
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Physical Form Liquid
CAS 886762-08-9
Health Hazard 1 Warning
Color Brown
Chemical Name or Material 5-Bromo-2-(trifluoromethoxy)aniline
UN Number UN2810
Quantity 25g
Assay 98%
Safety and Handling

Warning

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.