missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-pyrazinamine, Tech., Thermo Scientific™

Catalog Number p-3075778
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
59489-71-3
C4H4BrN3
174.001
MFCD00235015
KRRTXVSBTPCDOS-UHFFFAOYSA-N
2-amino-5-bromopyrazine, 5-bromo-2-pyrazinamine, 5-bromo-pyrazin-2-ylamine, 2-bromo-5-aminopyrazine, pyrazinamine, 5-bromo, 5-amino-2-bromopyrazine, 5-bromo-2-aminopyrazine, 5-bromopyrazin-2-ylamine, 5-bromopyrazinamine, 2-amino-5-bromo-pyrazine
599539
5-bromopyrazin-2-amine
C1=C(N=CC(=N1)Br)N
This item is not returnable. View return policy
59489-71-3
C4H4BrN3
174.001
MFCD00235015
KRRTXVSBTPCDOS-UHFFFAOYSA-N
2-amino-5-bromopyrazine, 5-bromo-2-pyrazinamine, 5-bromo-pyrazin-2-ylamine, 2-bromo-5-aminopyrazine, pyrazinamine, 5-bromo, 5-amino-2-bromopyrazine, 5-bromo-2-aminopyrazine, 5-bromopyrazin-2-ylamine, 5-bromopyrazinamine, 2-amino-5-bromo-pyrazine
599539
5-bromopyrazin-2-amine
C1=C(N=CC(=N1)Br)N
CAS Min % 97.0
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 174
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-Bromo-2-pyrazinamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'