missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-pyrazinamine, Tech., Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 97.0 |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 174 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 5-Bromo-2-pyrazinamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'