missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(methylthio)pyrimidine, 97%, ACROS Organics™

Brand:  Acros Organics 432380050

Code : 29

Additional Details : CAS Number : 14001-67-3 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10028199

  • £138.90 / 5g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

5-Bromo-2-(methylthio)pyrimidine
96.0
97%
C5H5BrN2S
MFCD00187875
14001-67-3
100.0
97%
205.08

Specifications

5-Bromo-2-(methylthio)pyrimidine
96.0
Authentic
64.0° to 68.0°C
Crystalline Powder of Flakes
97%
C5H5BrN2S
MFCD00187875
14001-67-3
100.0
Glass bottle
White
5g
97%
205.08
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Bromo-2-(methylthio)pyrimidine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS