missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methylaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-3212214
50.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
39478-78-9
MFCD00800678
RXQNKKRGJJRMKD-UHFFFAOYSA-N
2-amino-4-bromotoluene, 5-bromo-o-toluidine, 3-bromo-6-methylaniline, 5-bromo-2-methyl-aniline, benzenamine, 5-bromo-2-methyl, 5-bromo-2-methyl aniline, 5-bromo-2-methyl-phenylamine, zlchem 583, pubchem3801, acmc-1ajfs
2734805
5-bromo-2-methylaniline
CC1=C(C=C(C=C1)Br)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
39478-78-9
MFCD00800678
RXQNKKRGJJRMKD-UHFFFAOYSA-N
2-amino-4-bromotoluene, 5-bromo-o-toluidine, 3-bromo-6-methylaniline, 5-bromo-2-methyl-aniline, benzenamine, 5-bromo-2-methyl, 5-bromo-2-methyl aniline, 5-bromo-2-methyl-phenylamine, zlchem 583, pubchem3801, acmc-1ajfs
2734805
5-bromo-2-methylaniline
CC1=C(C=C(C=C1)Br)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.4900g/mL
Boiling Point 139°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H8BrN
Linear Formula BrC6H3(CH3)NH2
Refractive Index 1.6120 to 1.614
Quantity 25 g
Specific Gravity 1.49
Molecular Weight (g/mol) 186.05
Formula Weight 186.05
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 5-Bromo-2-methylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'