missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyrimidine, 97%, Thermo Scientific Chemicals

Catalog Number p-4258810
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14001-66-2
C5H5BrN2O
189.01
DWVCZDMMGYIULX-UHFFFAOYSA-N
2-methoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2-methoxy, 5-bromo-2-methoxy-pyrimidine, 5-bromo-2-methoxy-1,3-diazine, 5-bromopyrimidin-2-yl methyl ether, 5-bromo-2-methyloxy pyrimidine, pubchem5264, acmc-209cjt, 5-bromo-2methoxypyrimidine, ksc494g8n
139657
5-bromo-2-methoxypyrimidine
COC1=NC=C(C=N1)Br
This item is not returnable. View return policy
14001-66-2
C5H5BrN2O
189.01
DWVCZDMMGYIULX-UHFFFAOYSA-N
2-methoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2-methoxy, 5-bromo-2-methoxy-pyrimidine, 5-bromo-2-methoxy-1,3-diazine, 5-bromopyrimidin-2-yl methyl ether, 5-bromo-2-methyloxy pyrimidine, pubchem5264, acmc-209cjt, 5-bromo-2methoxypyrimidine, ksc494g8n
139657
5-bromo-2-methoxypyrimidine
COC1=NC=C(C=N1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 57°C to 59°C
Color Pink-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 189.01
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-methoxypyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-methoxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'