missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxypyridine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 11986371
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
103058-87-3
C7H6BrNO2
216.03
MFCD05663521
VRNOWEKCASDTFG-UHFFFAOYSA-N
5-bromo-2-methoxy-pyridine-3-carbaldehyde, 5-bromo-2-methoxynicotinaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxypyridin-3-carboxaldehyde, 5-bromo-2-methoxy-3-pyridinecarboxaldehyde, pubchem9207, abbypharma ap-10-5904, 3-pyridinecarboxaldehyde,5-bromo-2-methoxy, 5-bromo-2-methoxypyridine-3-carboxaldehyde 1g
1516403
5-bromo-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(Br)C=C1C=O
This item is not returnable. View return policy
103058-87-3
C7H6BrNO2
216.03
MFCD05663521
VRNOWEKCASDTFG-UHFFFAOYSA-N
5-bromo-2-methoxy-pyridine-3-carbaldehyde, 5-bromo-2-methoxynicotinaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxypyridin-3-carboxaldehyde, 5-bromo-2-methoxy-3-pyridinecarboxaldehyde, pubchem9207, abbypharma ap-10-5904, 3-pyridinecarboxaldehyde,5-bromo-2-methoxy, 5-bromo-2-methoxypyridine-3-carboxaldehyde 1g
1516403
5-bromo-2-methoxypyridine-3-carbaldehyde
COC1=NC=C(Br)C=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 102.0°C to 104.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 216.04
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 5-Bromo-2-methoxypyridine-3-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-methoxypyridine-3-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'