missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-hydroxy-4-methylpyridine, 97%

Brand:  Alfa Aesar™ H64674.14

Code : NEW

Additional Details : CAS Number : 164513-38-6 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15461979

  • £88.80 / 25g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Description

Description

Applications
It is used for medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

5-Bromo-2-hydroxy-4-methylpyridine
188.02
6798531
164513-38-6
MFCD03427664
5-Bromo-2-hydroxy-4-picoline

Specifications

5-Bromo-2-hydroxy-4-methylpyridine
Off-white
Solid
25g
MFCD03427664
5-Bromo-2-hydroxy-4-picoline
164513-38-6
198° to 202°C
97%
188.02
6798531
Videos
SDS