missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-hydroxy-3-methylpyridine, 98%

Brand:  Alfa Aesar™ H64253.06

Code : NEW

Additional Details : CAS Number : 89488-30-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15461359

  • £24.60 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Bromo-2-hydroxy-3-methylpyridine
188.02
1447443
89488-30-2
MFCD03427657
5-Bromo-2-hydroxy-3-picoline

Specifications

5-Bromo-2-hydroxy-3-methylpyridine
Off-white
Solid
5g
MFCD03427657
5-Bromo-2-hydroxy-3-picoline
89488-30-2
170° to 174°C (lit.)
98%
188.02
1447443
Videos
SDS