missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-hydroxy-3-methoxybenzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Linear Formula | CH3OC6H2(OH)(Br)CH(O) |
Quantity | 50 g |
Synonym | 5-bromo-3-methoxysalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy-3-methoxy, 5-bromo-2-vanillin, acmc-20am5q, 3-methoxy-5-bromosalicylaldehyde, 5bromo-2-hydroxy-3-methoxy-benzaldehyde, 5-bromo-2-hydroxy-3-methoxy-benzaldehyde, benzaldehyde,5-bromo-2-hydroxy-3-methoxy, 5-bromo-2-hydroxy-3-methoxybenzaldehyde |
Formula Weight | 231.04 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-2-hydroxy-3-methoxybenzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'