missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorotoluene, 99%, ACROS Organics™

Brand:  Acros Organics 269830050

Code : 29

Additional Details : CAS Number : 51437-00-4 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10573411

  • £8.20 / 5g


Estimated Shipment
13-07-2018
Add to basket


Specifications

Chemical Identifiers

5-Bromo-2-fluorotoluene
51437-00-4
100.0
99%
CH3C6H3BrF
MFCD00000343
99%
98.5
99%
C7H6BrF
189.03

Specifications

5-Bromo-2-fluorotoluene
51437-00-4
100.0
Glass bottle
1.5170g/mL
Liquid
94.0° to 95.0°C (50.0 mmHg)
5g
C7H6BrF
189.03
99%
98.5
Authentic
1.5280 to 1.5300
Clear colorless
99%
99%
73°C
CH3C6H3BrF
MFCD00000343
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Bromo-2-fluorotoluene

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS