missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-fluoropyridine-3-carboxaldehyde, 95%

Brand:  Alfa Aesar™ H61123.03

Code : NEW

Additional Details : CAS Number : 875781-15-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447628

  • £65.00 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Bromo-2-fluoropyridine-3-carboxaldehyde
204
14515665
875781-15-0
MFCD09842440
5-Bromo-2-fluoronicotinaldehyde

Specifications

5-Bromo-2-fluoropyridine-3-carboxaldehyde
Off-white to pale yellow
95%
204
14515665
875781-15-0
Powder
1g
MFCD09842440
5-Bromo-2-fluoronicotinaldehyde
Videos
SDS