missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzoic acid, 97%, ACROS Organics™

£19.79 - £66.91

Chemical Identifiers

CAS 146328-85-0
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219
MDL Number MFCD00143423
InChI Key PEXAZYDITWXYNJ-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromobenzoic acid, 5-bromo-2-fluoro-benzoic acid, buttpark 19\01-67, benzoic acid, 5-bromo-2-fluoro, 3-bromo-6-fluoro-benzoic acid, pubchem1312, acmc-209cww, 3-bromo-6-fluorobenzoic acid, ksc183q6j, rarechem al bo 0757
PubChem CID 2736315
IUPAC Name 5-bromo-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10001074
SDS View Documents
Acros Organics
398590010
1g Glass bottle
£19.79
1g
View alternative products
Save Up to 
»
Estimated Shipment
11-12-2020
Please to purchase this item. Need a web account? Register with us today!
10604175
SDS View Documents
Acros Organics
398590050
5g Glass bottle
£66.91
5g
View alternative products
Save Up to 
»
Estimated Shipment
06-01-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

146328-85-0
219
PEXAZYDITWXYNJ-UHFFFAOYSA-N
2736315
C1=CC(=C(C=C1Br)C(=O)O)F
C7H4BrFO2
MFCD00143423
2-fluoro-5-bromobenzoic acid, 5-bromo-2-fluoro-benzoic acid, buttpark 19\01-67, benzoic acid, 5-bromo-2-fluoro, 3-bromo-6-fluoro-benzoic acid, pubchem1312, acmc-209cww, 3-bromo-6-fluorobenzoic acid, ksc183q6j, rarechem al bo 0757
5-bromo-2-fluorobenzoic acid

Specifications

5-Bromo-2-fluorobenzoic acid
141°C to 145°C
146328-85-0
100.0
C7H4BrFO2
MFCD00143423
PEXAZYDITWXYNJ-UHFFFAOYSA-N
5-bromo-2-fluorobenzoic acid
2736315
97%
Glass bottle
1g
96.0
96% min. (HPLC)
BrC6H3(F)COOH
2-fluoro-5-bromobenzoic acid, 5-bromo-2-fluoro-benzoic acid, buttpark 19\01-67, benzoic acid, 5-bromo-2-fluoro, 3-bromo-6-fluoro-benzoic acid, pubchem1312, acmc-209cww, 3-bromo-6-fluorobenzoic acid, ksc183q6j, rarechem al bo 0757
C1=CC(=C(C=C1Br)C(=O)O)F
219
219
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Bromo-2-fluorobenzoic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS